Learn More
Bellco Glass, Inc. MicroCarrier Spinner Flask complete,3L
SDP
Catalog No. 501944973
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 3l Center Cap:100mm Sidearm cap: 45mm OD X OAH:165 X 280
Catalog No. 50-194-4973 Supplier Bellco Glass, Inc. Supplier No. 196503000
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 3l Center Cap:100mm Sidearm cap: 45mm OD X OAH:165 X 280