Learn More
Bellco Glass, Inc. MicroCarrier Spinner Flask Only, 250mL W/Dimple Bottom - No Ctr Cap
SDP
Catalog No. 501944978
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 250ml Sidearm cap: 32mm (included)
Catalog No. 50-194-4978 Supplier Bellco Glass, Inc. Supplier No. 196510250
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 250ml Sidearm cap: 32mm (included)