Learn More
Bellco Glass, Inc. MicroCarrier Spinner Flask Only, 3L No Ctr Cap
SDP
Catalog No. 501944983
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 3l Sidearm cap: 45mm (included)
Catalog No. 50-194-4983 Supplier Bellco Glass, Inc. Supplier No. 196513000
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 3l Sidearm cap: 45mm (included)