Learn More
Chemglass Life Sciences Spinner Flask, 3000mL, Flat Bottom, Replacement Glass Flask ONLY
SDP
Catalog No. 501214633
Encompass_Preferred
Replacement vessel is supplied with black phenolic, rubber lined caps for the 18mm side arms, polypropylene caps and pouring rings for the GL-32 and GL-45 side arms.
Catalog No. 50-121-4633 Supplier Chemglass Life Sciences Supplier No. CLS14003L
Replacement vessel is supplied with black phenolic, rubber lined caps for the 18mm side arms, polypropylene caps and pouring rings for the GL-32 and GL-45 side arms.