Learn More
Bellco Glass, Inc. MicroCarrier Spinner complete,500mL,100mm Neck
SDP
Catalog No. 501944972
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 500ml Center Cap:100mm Sidearm cap: 45mm OD X OAH:110 X 180
Catalog No. 50-194-4972 Supplier Bellco Glass, Inc. Supplier No. 196502500
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 500ml Center Cap:100mm Sidearm cap: 45mm OD X OAH:110 X 180