Learn More
Bellco Glass, Inc. MicroCarrier Spinner Flask complete, 100mL
SDP
Catalog No. 501944967
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 100ml Center Cap:70mm Sidearm cap: 32mm OD X OAH:65 X 135
Catalog No. 50-194-4967 Supplier Bellco Glass, Inc. Supplier No. 196500100
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 100ml Center Cap:70mm Sidearm cap: 32mm OD X OAH:65 X 135