Learn More
Bellco Glass, Inc. MicroCarrier Spinner Flask Complete,15L
SDP
Catalog No. 501944986
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 15l Sidearm cap: 45mm Center cap: 100mm OD X OAH:294 X 470 (with cap)
Catalog No. 50-194-4986 Supplier Bellco Glass, Inc. Supplier No. 196515000
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 15l Sidearm cap: 45mm Center cap: 100mm OD X OAH:294 X 470 (with cap)