Learn More
Bellco Glass, Inc. MicroCarrier Spinner Flask complete, 250mL
SDP
Catalog No. 501944968
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 250ml Center Cap:70mm Sidearm cap: 32mm OD X OAH:85 X 140
Catalog No. 50-194-4968 Supplier Bellco Glass, Inc. Supplier No. 196500250
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 250ml Center Cap:70mm Sidearm cap: 32mm OD X OAH:85 X 140