Learn More
Bellco Glass, Inc. MicroCarrier Spinner Flask complete,6L
SDP
Catalog No. 501944975
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 6l Center Cap:100mm Sidearm cap: 45mm OD X OAH:204 X 410
Catalog No. 50-194-4975 Supplier Bellco Glass, Inc. Supplier No. 196506000
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 6l Center Cap:100mm Sidearm cap: 45mm OD X OAH:204 X 410