Learn More
Bellco Glass, Inc. MicroCarrier Spinner Flask complete,8L
SDP
Catalog No. 501944976
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 8l Center Cap:100mm Sidearm cap: 45mm OD X OAH:237 X 430
Catalog No. 50-194-4976 Supplier Bellco Glass, Inc. Supplier No. 196508000
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 8l Center Cap:100mm Sidearm cap: 45mm OD X OAH:237 X 430