Learn More
Bellco Glass, Inc. MicroCarrier Spinner Flask Only, 15L No Ctr Cap
SDP
Catalog No. 501944982
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 15l Sidearm cap: 45mm (included)
Catalog No. 50-194-4982 Supplier Bellco Glass, Inc. Supplier No. 196511500
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 15l Sidearm cap: 45mm (included)