Learn More
Bellco Glass, Inc. MicroCarrier Spinner Flask Only, 1L W/Dimple Bottom - No Ctr Cap
SDP
Catalog No. 501944980
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 1l Sidearm cap: 45mm (included)
Catalog No. 50-194-4980 Supplier Bellco Glass, Inc. Supplier No. 196511000
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 1l Sidearm cap: 45mm (included)