Learn More
Bellco Glass, Inc. MicroCarrier Spinner Flask Only, 36L No Ctr Cap
SDP
Catalog No. 501944985
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. OD x oah:410mm x 535mm Flask Only
Catalog No. 50-194-4985 Supplier Bellco Glass, Inc. Supplier No. 196513600
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. OD x oah:410mm x 535mm Flask Only