Learn More
Bellco Glass, Inc. MicroCarrier Spinner Flask Only, 6L No Ctr Cap
SDP
Catalog No. 501944987
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 6l Sidearm cap: 45mm (included) •Material: Type 1 Class A Boro Glass•100mm Center Neck•Two (2) angled 45mm sidearms (30°)•Dimensions : 204mm OD X 410mm OAH•Grad. Range: 1L - 6L, every 1L•Supply with 45mm PP screw caps & pouring rings
Catalog No. 50-194-4987 Supplier Bellco Glass, Inc. Supplier No. 196516000
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 6l Sidearm cap: 45mm (included) •Material: Type 1 Class A Boro Glass•100mm Center Neck•Two (2) angled 45mm sidearms (30°)•Dimensions : 204mm OD X 410mm OAH•Grad. Range: 1L - 6L, every 1L•Supply with 45mm PP screw caps & pouring rings