Learn More
Zyagen Labs Mouse C57 liver total RNA is a high pure intact total RNA isolated from freshly harvested normal healthy C57BL/6 Mouse LIVER tissues, treated with DNase and dissolved in water. RNA is ready for immediate use in any downstream application.
SDP
Catalog No. 501718332
Encompass_Preferred
Zyagen Total RNA is routinely extracted from freshly harvested normal healthy tissues of single donor using classical guanidine isothiocyanate–phenol:chloroform extraction method which allows the rapid isolation of total RNA including microRNAs. RNA is treated with RNase-free DNase to remove residual DNA, precisely quantified, and stored at -80oC. The integrity of each RNA sample as indicated by intact ribosomal RNA is verified by denatured agarose gel electrophoresis. The purity of RNA is assessed by NanoDrop (A260/A280: 1.9-2.1). Residual DNA contamination is tested by PCR. RNA is ideal for Northern blotting, ribonuclease protection assay, SI nuclease assay, RT-PCR/Q-PCR/RACE analysis, cDNA synthesis, RNA differential display, microRNA studies, and purification of mRNA for library construction. Total RNA sample is provided in RNase-free water at a concentration of 1 mg/ml and shipped on dry ice.
Catalog No. 50-171-8332 Supplier Zyagen Labs Supplier No. MR314C57
Zyagen Total RNA is routinely extracted from freshly harvested normal healthy tissues of single donor using classical guanidine isothiocyanate–phenol:chloroform extraction method which allows the rapid isolation of total RNA including microRNAs. RNA is treated with RNase-free DNase to remove residual DNA, precisely quantified, and stored at -80oC. The integrity of each RNA sample as indicated by intact ribosomal RNA is verified by denatured agarose gel electrophoresis. The purity of RNA is assessed by NanoDrop (A260/A280: 1.9-2.1). Residual DNA contamination is tested by PCR. RNA is ideal for Northern blotting, ribonuclease protection assay, SI nuclease assay, RT-PCR/Q-PCR/RACE analysis, cDNA synthesis, RNA differential display, microRNA studies, and purification of mRNA for library construction. Total RNA sample is provided in RNase-free water at a concentration of 1 mg/ml and shipped on dry ice.