Learn More
Medchemexpress LLC M-carboxycinnamic acid bishydroxamide | 174664-65-4 | MFCD03453553 | 98.0% | 222.20 g·mol⁻¹ | C10H10N2O4 | 10 MG
SDP
Catalog No. 5000304172
Encompass_Preferred
m-Carboxycinnamic acid bishydroxamide (CBHA) is a small-molecule histone deacetylase (HDAC) inhibitor supplied for research use. It inhibits HDAC1 and HDAC3 with reported ID50 values of approximately 10 nM and 70 nM, respectively, and has been reported to induce apoptosis and suppress tumor growth in experimental models.
  • Potent HDAC inhibitor with reported ID50 ≈ 10 nM (HDAC1) and ≈ 70 nM (HDAC3).
  • High purity (98.0%).
  • Molecular formula C10H10N2O4 and molecular weight 222.20 g·mol⁻¹.
  • Suitable for biochemical and cell-based assays.
  • Demonstrated induction of apoptosis and tumor-growth suppression in studies.
  • Supplied as a research-grade chemical for laboratory use.
Catalog No. 50-003-04172 Supplier Medchemexpress LLC Supplier No. HYW01400410MG
m-Carboxycinnamic acid bishydroxamide (CBHA) is a small-molecule histone deacetylase (HDAC) inhibitor supplied for research use. It inhibits HDAC1 and HDAC3 with reported ID50 values of approximately 10 nM and 70 nM, respectively, and has been reported to induce apoptosis and suppress tumor growth in experimental models.
  • Potent HDAC inhibitor with reported ID50 ≈ 10 nM (HDAC1) and ≈ 70 nM (HDAC3).
  • High purity (98.0%).
  • Molecular formula C10H10N2O4 and molecular weight 222.20 g·mol⁻¹.
  • Suitable for biochemical and cell-based assays.
  • Demonstrated induction of apoptosis and tumor-growth suppression in studies.
  • Supplied as a research-grade chemical for laboratory use.