Learn More
Andwin Scientific M-CELL 6/PK
SDP
Catalog No. NC1052692
Encompass
The M-Cell or Six Pack is composed of six test kits. Each test kit has vacuum packed expendables that are used to conduct one test. This product also contains a container of distilled water to perform water separation tests.
Catalog No. NC1052692 Supplier Andwin Scientific Supplier No. 840995946
The M-Cell or Six Pack is composed of six test kits. Each test kit has vacuum packed expendables that are used to conduct one test. This product also contains a container of distilled water to perform water separation tests.