Learn More
Lifeline Cell Technology M CORNEPI HU LIFELINE KT
SDP
Catalog No. 50311903
Encompass_Preferred
OcuLife is a new medium optimized for the culture of human corneal epithelial cells. OcuLife supports the growth of these cells over a period of at least three passages. OcuLife contains no antimicrobials and no phenol red
Catalog No. 50-311-903 Supplier Lifeline Cell Technology Supplier No. LL0032
OcuLife is a new medium optimized for the culture of human corneal epithelial cells. OcuLife supports the growth of these cells over a period of at least three passages. OcuLife contains no antimicrobials and no phenol red