Learn More
Cayman Chemical m-Coumaric Acid, Purity: ≥98%, CAS: 588-30-7, 10 g
SDP
Catalog No. 502915359
Encompass_Preferred
An active metabolite of polyphenols; inhibits cumene hydroperoxide-induced oxidation of DNA (IC50 = 54.7 µM); induces neurite outgrowth of fetal rat hippocampal neurons at 10 µM; induces apoptosis and cell cycle arrest at the G0/G1 phase in HeLa cells at 5 and 10 mM, respectively; increases locomotor activity in mice at 3 mmol/kg
Catalog No. 50-291-5359 Supplier Cayman Chemical Supplier No. 3723610 G
An active metabolite of polyphenols; inhibits cumene hydroperoxide-induced oxidation of DNA (IC50 = 54.7 µM); induces neurite outgrowth of fetal rat hippocampal neurons at 10 µM; induces apoptosis and cell cycle arrest at the G0/G1 phase in HeLa cells at 5 and 10 mM, respectively; increases locomotor activity in mice at 3 mmol/kg