Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Cresol Purple, 0.1% Aqueous, pH 1.2 to 2.8 Red to Yellow, pH 7.4 to 9.0 Yellow to Purple, Certified, LabChem™

Catalog No. LC135001 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC135001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC135001 Supplier LabChem, Inc. Supplier No. LC135001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified indicator for laboratory application.

Chemical Identifiers

CAS 62625-31-4 , 7732-18-5
Molecular Weight (g/mol) 404.42

Specifications

CAS Max % 99.9, 0.1
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Linear Formula C21H17O5SNa
Decomposition Information Carbon monoxide; Carbon dioxide; Sulfur compounds
Density 1g/mL
Color Purple/Red
Solubility Information Soluble in water
Quantity 500 mL
Physical Form Liquid
Formula Weight 404.42
Grade Certified
Chemical Name or Material m-Cresol Purple, 0.1% Aqueous, pH 1.2 to 2.8 Red to Yellow, pH 7.4 to 9.0 Yellow to Purple
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.