Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Cresol Purple Indicator, 0.04% (w/v), Ricca Chemical
SDP

Catalog No. R24000001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R24000001A 1 L Natural Poly Bottle
2400-4 120 mL Natural Poly Bottle
2400-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R24000001A Supplier Ricca Chemical Company Supplier No. 240032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 62625-31-4 , 7732-18-5

Specifications

CAS Min % 98.0, 0.04
CAS Max % 100.0, 0.04
Packaging Natural Poly Bottle
Color Red/Orange
pH 7 to 9
Quantity 1 L
Physical Form Liquid
Percent Purity 0.04% W/V
Grade Indicator
Chemical Name or Material m-Cresol Purple Indicator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.