Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals m-Cresol Purple, pure, indicator grade

Catalog No. AC228200100
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC228200100 10 g Glass bottle
AC228200050 5 g Glass bottle
AC228200500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC228200100 Supplier Thermo Scientific Chemicals Supplier No. 228200100
Add to Cart

CAS: 2303-01-7 | C21H18O5S | 382.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2303-01-7
Molecular Formula C21H18O5S
Molecular Weight (g/mol) 382.43
MDL Number MFCD00005871
InChI Key OLQIKGSZDTXODA-UHFFFAOYSA-N
PubChem CID 73030
IUPAC Name 4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
SMILES CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C

Specifications

Infrared Spectrum Authentic
Formula Weight 382.42
Grade Pure
Packaging Glass bottle
Specific Extinction 900 min. (578nm, in buffer)
Visual Transition Interval from pH 7.4 (yellow) to pH 9.0 (purple)
Color Green
Quantity 10 g
Physical Form Fine Crystalline Powder
Chemical Name or Material m-Cresol Purple
EINECSNumber 218-960-6
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.