Learn More
Sigma Aldrich Fine Chemicals Biosciences m-Cresol Purple sodium sal10G
Catalog No. 502898666
Encompass
m-cresol purple sodium salt (MCP) is a colorimetric pH indicator dye. The dye is blue in anionic form and yellow in protonated form.
Catalog No. 50-289-8666 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 21176110G
m-cresol purple sodium salt (MCP) is a colorimetric pH indicator dye. The dye is blue in anionic form and yellow in protonated form.