Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Cresol Purple Sodium Salt, Ricca Chemical
SDP

Catalog No. RDCC080010F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCC080010F 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCC080010F Supplier Ricca Chemical Company Supplier No. RDCC080010F1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 62625-31-4
Molecular Weight (g/mol) 404.41 g/mol

Specifications

CAS Min % 98.0
CAS Max % 100.0
Packaging Pouch
Color Black
Quantity 10 g
Physical Form Metallic Powder
Grade Indicator
Chemical Name or Material m-Cresol Purple Sodium Salt
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.