Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Cresol Purple Sodium Salt, Sigma-Aldrich

Catalog No. 01258848 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-848 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-848 Supplier MilliporeSigma Supelco Supplier No. 21176110G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Colorimetric pH indicator dye used for spectrophotometric measurements of seawater pH and diagnostic assay manufacturing.

m-Cresol Purple sodium salt has been used to measure pH of seawater using a spectrophotometer.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-31-4
Molecular Weight (g/mol) 404.41 g/mol

Specifications

Concentration 90%
Color Black
Quantity 10 g
Grade Spectrophotometry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.