Learn More
Medchemexpress LLC HY-P7085A 10ug Medchemexpress, M-CSF (CHO-expressed), Mouse CAS: Purity:>98%
SDP
Catalog No. 501934325
Encompass_Preferred
Medchemexpress, HY-P7085A 10ug M-CSF (CHO-expressed), Mouse CAS: Recombinant Mouse Macrophage Colony Stimulating Factor (CHO-expressed) is a pro-inflammatory cytokine, binds to its receptor CSF1R, and is involved in the development and proliferation of cells of the monocyte/macrophage lineage and participates in the induction of osteoclasts which are important in the destruction of bone and cartilage and in the periarticular osteoporotic changes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4325 Supplier Medchemexpress LLC Supplier No. HYP7085A10UG
Medchemexpress, HY-P7085A 10ug M-CSF (CHO-expressed), Mouse CAS: Recombinant Mouse Macrophage Colony Stimulating Factor (CHO-expressed) is a pro-inflammatory cytokine, binds to its receptor CSF1R, and is involved in the development and proliferation of cells of the monocyte/macrophage lineage and participates in the induction of osteoclasts which are important in the destruction of bone and cartilage and in the periarticular osteoporotic changes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.