Learn More
Medchemexpress LLC HY-P7247 10ug Medchemexpress, M-CSF (CHO-expressed), Rat CAS: Purity:>98%
SDP
Catalog No. 501934369
Encompass_Preferred
Medchemexpress, HY-P7247 10ug M-CSF (CHO-expressed), Rat CAS: Recombinant Rat Macrophage-Colony Stimulating Factor (CHO-expressed) is a pro-inflammatory cytokine which binds to its receptor CSF1R, and is involved in the development and proliferation of cells of the monocyte/macrophage lineage and participates in the induction of osteoclasts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4369 Supplier Medchemexpress LLC Supplier No. HYP724710UG
Medchemexpress, HY-P7247 10ug M-CSF (CHO-expressed), Rat CAS: Recombinant Rat Macrophage-Colony Stimulating Factor (CHO-expressed) is a pro-inflammatory cytokine which binds to its receptor CSF1R, and is involved in the development and proliferation of cells of the monocyte/macrophage lineage and participates in the induction of osteoclasts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.