Learn More
Medchemexpress LLC M-CSF Human 10ug
SDP
Catalog No. 5000185008
Encompass_Preferred
Recombinant human macrophage colony-stimulating factor (M-CSF, CSF-1) is supplied as a lyophilized, tag-free protein for in vitro research. It promotes proliferation and differentiation of mononuclear phagocyte lineage cells and is provided with confirmed purity and specific biological activity for cell-based assays and biochemical studies.
  • Homodimeric hematopoietic growth factor that stimulates mononuclear phagocyte proliferation.
  • Lyophilized, tag-free recombinant protein for flexible reconstitution.
  • Expressed in E. coli to provide consistent material for assays.
  • Greater than 95% purity as determined by reducing SDS-PAGE.
  • High biological potency with ED50 in the low ng/mL range in M-NFS-60 proliferation assay.
  • Available in small research sizes suitable for pilot studies and optimization.
  • Stable when stored at -20°C; aliquot and freeze after reconstitution for long-term storage.
Catalog No. 50-001-85008 Supplier Medchemexpress LLC Supplier No. HYP705010UG
Recombinant human macrophage colony-stimulating factor (M-CSF, CSF-1) is supplied as a lyophilized, tag-free protein for in vitro research. It promotes proliferation and differentiation of mononuclear phagocyte lineage cells and is provided with confirmed purity and specific biological activity for cell-based assays and biochemical studies.
  • Homodimeric hematopoietic growth factor that stimulates mononuclear phagocyte proliferation.
  • Lyophilized, tag-free recombinant protein for flexible reconstitution.
  • Expressed in E. coli to provide consistent material for assays.
  • Greater than 95% purity as determined by reducing SDS-PAGE.
  • High biological potency with ED50 in the low ng/mL range in M-NFS-60 proliferation assay.
  • Available in small research sizes suitable for pilot studies and optimization.
  • Stable when stored at -20°C; aliquot and freeze after reconstitution for long-term storage.