Learn More
Medchemexpress LLC M-CSF Human 223a.a 100ug
SDP
Catalog No. 5000184675
Encompass_Preferred
Recombinant human macrophage colony-stimulating factor 1 (M-CSF) is a 223-amino-acid cytokine produced in HEK293 cells with a C-terminal 6xHis tag. The lyophilized protein is >95% pure and appears at ~40-50 kDa due to glycosylation; it is supplied with validated biological activity for use in macrophage and monocyte proliferation, differentiation, and signaling studies.
  • Expressed in human HEK293 cells for native glycosylation.
  • C-terminal 6xHis tag enables detection and purification.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Measured biological activity with low ED50 for reliable assays.
  • Lyophilized formulation for room-temperature shipping and long-term storage at -20°C.
  • Multiple unit sizes available to suit experimental scale.
Catalog No. 50-001-84675 Supplier Medchemexpress LLC Supplier No. HYP70488100UG
Recombinant human macrophage colony-stimulating factor 1 (M-CSF) is a 223-amino-acid cytokine produced in HEK293 cells with a C-terminal 6xHis tag. The lyophilized protein is >95% pure and appears at ~40-50 kDa due to glycosylation; it is supplied with validated biological activity for use in macrophage and monocyte proliferation, differentiation, and signaling studies.
  • Expressed in human HEK293 cells for native glycosylation.
  • C-terminal 6xHis tag enables detection and purification.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Measured biological activity with low ED50 for reliable assays.
  • Lyophilized formulation for room-temperature shipping and long-term storage at -20°C.
  • Multiple unit sizes available to suit experimental scale.