Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-CSF Human 223a.a 2ug
SDP

Catalog No. 5000185009
Encompass_Preferred

Recombinant human macrophage colony-stimulating factor (M-CSF) expressed in HEK293 cells with a C-terminal 6xHis tag and supplied lyophilized for research use. The protein supports macrophage survival, proliferation, and differentiation, and is provided with high purity plus recommended storage and reconstitution guidance to help maintain activity.

  • Recombinant human M-CSF, 223 amino acid sequence.
  • Expressed in HEK293 cells with a C-terminal 6xHis tag.
  • Lyophilized formulation for improved stability and transport.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Predicted molecular mass 26.6 kDa; glycosylated form appears ~40-50 kDa on SDS-PAGE.
  • Reconstitution guidance: do not reconstitute below 100 μg/mL; aliquot and store at -20°C or -80°C.
  • For research use only.

Catalog No. 50-001-85009 Supplier Medchemexpress LLC Supplier No. HYP704882UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human macrophage colony-stimulating factor (M-CSF) expressed in HEK293 cells with a C-terminal 6xHis tag and supplied lyophilized for research use. The protein supports macrophage survival, proliferation, and differentiation, and is provided with high purity plus recommended storage and reconstitution guidance to help maintain activity.

  • Recombinant human M-CSF, 223 amino acid sequence.
  • Expressed in HEK293 cells with a C-terminal 6xHis tag.
  • Lyophilized formulation for improved stability and transport.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Predicted molecular mass 26.6 kDa; glycosylated form appears ~40-50 kDa on SDS-PAGE.
  • Reconstitution guidance: do not reconstitute below 100 μg/mL; aliquot and store at -20°C or -80°C.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.