Learn More
Sigma Aldrich Fine Chemicals Biosciences M CSf human Animal compone
Catalog No. 502905333
Encompass
M-CSf (macrophage colony-stimulating factor) is a potent hematopoietic factor produced by a variety of cells including lymphocytes monocytes fibroblasts endothelial cells myoblasts and osteoblasts. The protein works through its receptor CSf1R (colony stimulating factor 1 receptor). The human is reactive in murine systems but the murine molecule exhibits no activity on human cells. Recombinant human M-CSf is a 36.8kDa homodimeric protein containing two 159 amino acid polypeptide subunits.
Catalog No. 50-290-5333 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRP311010UG
M-CSf (macrophage colony-stimulating factor) is a potent hematopoietic factor produced by a variety of cells including lymphocytes monocytes fibroblasts endothelial cells myoblasts and osteoblasts. The protein works through its receptor CSf1R (colony stimulating factor 1 receptor). The human is reactive in murine systems but the murine molecule exhibits no activity on human cells. Recombinant human M-CSf is a 36.8kDa homodimeric protein containing two 159 amino acid polypeptide subunits.