Learn More
Medchemexpress LLC M-CSF Human CHO 100ug
SDP
Catalog No. 5000185042
Encompass_Preferred
Recombinant human macrophage colony-stimulating factor (M-CSF) produced in CHO cells and supplied as a lyophilized protein for research applications. The glycosylated homodimer supports macrophage survival, proliferation, and differentiation in cell-based assays; product information includes sequence, bioactivity, purity, and recommended storage to preserve activity.
  • Glycosylated, CHO-expressed human protein with native post-translational modifications.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Verified bioactivity with ED50 in M-NFS-60 proliferation assay.
  • Lyophilized formulation dialyzed against PBS, pH 7.4, with trehalose for stability.
  • Recommended aliquoting and frozen storage to maintain long-term activity.
  • Available in multiple convenient sizes, including a 100 μg format.
Catalog No. 50-001-85042 Supplier Medchemexpress LLC Supplier No. HYP7050A100UG
Recombinant human macrophage colony-stimulating factor (M-CSF) produced in CHO cells and supplied as a lyophilized protein for research applications. The glycosylated homodimer supports macrophage survival, proliferation, and differentiation in cell-based assays; product information includes sequence, bioactivity, purity, and recommended storage to preserve activity.
  • Glycosylated, CHO-expressed human protein with native post-translational modifications.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Verified bioactivity with ED50 in M-NFS-60 proliferation assay.
  • Lyophilized formulation dialyzed against PBS, pH 7.4, with trehalose for stability.
  • Recommended aliquoting and frozen storage to maintain long-term activity.
  • Available in multiple convenient sizes, including a 100 μg format.