Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-CSF Human CHO 500ug
SDP

Catalog No. 5000184153
Encompass_Preferred

Recombinant human M-CSF (CSF-1), produced in CHO cells and supplied as a tag-free, lyophilized protein, is a glycosylated, disulfide-linked homodimer that promotes proliferation, differentiation, and polarization of monocyte-macrophage lineage cells. It is suitable for macrophage and osteoclast differentiation assays, signaling studies, and functional bioassays requiring biologically active human M-CSF.

  • CHO-expressed with mammalian glycosylation for native activity.
  • Tag free to avoid interference in binding and bioassays.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Apparent molecular weight reflects glycosylation (18-22 kDa reduced).
  • Lyophilized formulation for extended storage and convenient reconstitution.
  • Datasheet and certificate of analysis available for quality assurance.

Catalog No. 50-001-84153 Supplier Medchemexpress LLC Supplier No. HYP7050A500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human M-CSF (CSF-1), produced in CHO cells and supplied as a tag-free, lyophilized protein, is a glycosylated, disulfide-linked homodimer that promotes proliferation, differentiation, and polarization of monocyte-macrophage lineage cells. It is suitable for macrophage and osteoclast differentiation assays, signaling studies, and functional bioassays requiring biologically active human M-CSF.

  • CHO-expressed with mammalian glycosylation for native activity.
  • Tag free to avoid interference in binding and bioassays.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Apparent molecular weight reflects glycosylation (18-22 kDa reduced).
  • Lyophilized formulation for extended storage and convenient reconstitution.
  • Datasheet and certificate of analysis available for quality assurance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.