Learn More
Medchemexpress LLC M-CSF Human HEK293 10ug
SDP
Catalog No. 5000179169
Encompass_Preferred
The M-CSF protein is an important cytokine that modulates hematopoietic precursor cells especially mononuclear phagocytes affecting innate immunity and inflammation through the release of proinflammatory chemokines It is essential for osteoclast proliferation and regulates bone resorption bone development and fertility M-CSF Protein Human (HEK293 His) is the recombinant human-derived M-CSF protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-79169 Supplier Medchemexpress LLC Supplier No. HYP7382710UG
The M-CSF protein is an important cytokine that modulates hematopoietic precursor cells especially mononuclear phagocytes affecting innate immunity and inflammation through the release of proinflammatory chemokines It is essential for osteoclast proliferation and regulates bone resorption bone development and fertility M-CSF Protein Human (HEK293 His) is the recombinant human-derived M-CSF protein expressed by HEK293 with C-10 His labeled tag