Learn More
Medchemexpress LLC M-CSF Human HEK293 50ug
SDP
Catalog No. 5000179737
Encompass_Preferred
The M-CSF protein is an important cytokine that modulates hematopoietic precursor cells especially mononuclear phagocytes affecting innate immunity and inflammation through the release of proinflammatory chemokines It is essential for osteoclast proliferation and regulates bone resorption bone development and fertility M-CSF Protein Human (HEK293 Fc) is the recombinant human-derived M-CSF protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-79737 Supplier Medchemexpress LLC Supplier No. HYP7382950UG
The M-CSF protein is an important cytokine that modulates hematopoietic precursor cells especially mononuclear phagocytes affecting innate immunity and inflammation through the release of proinflammatory chemokines It is essential for osteoclast proliferation and regulates bone resorption bone development and fertility M-CSF Protein Human (HEK293 Fc) is the recombinant human-derived M-CSF protein expressed by HEK293 with C-hFc labeled tag