Learn More
Medchemexpress LLC M-CSF Mouse HEK293 10ug
SDP
Catalog No. 5000185377
Encompass_Preferred
Recombinant mouse macrophage colony-stimulating factor (M-CSF) expressed in HEK293 cells and supplied lyophilized for research use. The protein is biologically active in cell proliferation assays (ED50 ≤15 ng/mL), is glycosylated (apparent 32-60 kDa by SDS-PAGE), and has low endotoxin (<1 EU/μg). Lyophilized from PBS pH 7.4 (some lots with 8% trehalose); store at -20°C and reconstitute at ≥100 μg/mL in ddH2O with carrier protein for long-term storage.
  • Expressed in HEK293 mammalian cells for native glycosylation.
  • Validated bioactivity with ED50 ≤15 ng/mL in M-NFS-60 assay.
  • Low endotoxin level (<1 EU/μg) suitable for cell-based assays.
  • Lyophilized formulation enables convenient storage and transport.
  • Recommended reconstitution ≥100 μg/mL and aliquot storage at -20°C or -80°C.
  • Lot-specific buffer and COA information available on request.
Catalog No. 50-001-85377 Supplier Medchemexpress LLC Supplier No. HYP7055310UG
Recombinant mouse macrophage colony-stimulating factor (M-CSF) expressed in HEK293 cells and supplied lyophilized for research use. The protein is biologically active in cell proliferation assays (ED50 ≤15 ng/mL), is glycosylated (apparent 32-60 kDa by SDS-PAGE), and has low endotoxin (<1 EU/μg). Lyophilized from PBS pH 7.4 (some lots with 8% trehalose); store at -20°C and reconstitute at ≥100 μg/mL in ddH2O with carrier protein for long-term storage.
  • Expressed in HEK293 mammalian cells for native glycosylation.
  • Validated bioactivity with ED50 ≤15 ng/mL in M-NFS-60 assay.
  • Low endotoxin level (<1 EU/μg) suitable for cell-based assays.
  • Lyophilized formulation enables convenient storage and transport.
  • Recommended reconstitution ≥100 μg/mL and aliquot storage at -20°C or -80°C.
  • Lot-specific buffer and COA information available on request.