Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-CSF Mouse HEK293 500ug
SDP

Catalog No. 5000184544
Encompass_Preferred

Recombinant mouse macrophage colony-stimulating factor (M-CSF) expressed in HEK293 cells and supplied lyophilized. The product is glycosylated, tag free, and >95% pure by reducing SDS-PAGE, and is suitable for cell culture and biochemical assays requiring biologically active M-CSF.

  • Produced in HEK293 mammalian expression system.
  • Tag free and glycosylated recombinant protein.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized from 0.2 μm filtered PBS, pH 7.4, or PBS with 8% trehalose.
  • Biological activity: ED50 ≤15 ng/mL in M-NFS-60 cell proliferation assay.
  • Do not reconstitute below 100 μg/mL; aliquot and store at -20°C or -80°C for long-term storage.

Catalog No. 50-001-84544 Supplier Medchemexpress LLC Supplier No. HYP70553500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse macrophage colony-stimulating factor (M-CSF) expressed in HEK293 cells and supplied lyophilized. The product is glycosylated, tag free, and >95% pure by reducing SDS-PAGE, and is suitable for cell culture and biochemical assays requiring biologically active M-CSF.

  • Produced in HEK293 mammalian expression system.
  • Tag free and glycosylated recombinant protein.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized from 0.2 μm filtered PBS, pH 7.4, or PBS with 8% trehalose.
  • Biological activity: ED50 ≤15 ng/mL in M-NFS-60 cell proliferation assay.
  • Do not reconstitute below 100 μg/mL; aliquot and store at -20°C or -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.