Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-CSF protein, mouse (HEK293, C-His) | 81627-83-0 | >95.0% | 50UG
SDP

Supplier:  Medchemexpress LLC HYP70263A50UG

Encompass_Preferred

Recombinant mouse macrophage colony-stimulating factor (M-CSF) expressed in HEK293 cells with a C-terminal 6xHis tag. Supplied lyophilized and formulated for stability, it is intended for cell-based assays measuring proliferation, survival, and differentiation of myeloid-lineage cells.

  • Expressed in HEK293 cells.
  • C-terminal 6xHis tag for detection and purification.
  • Predicted molecular mass 26 kDa; observed ~32-60 kDa by SDS-PAGE due to glycosylation.
  • Purity >95% by reducing SDS-PAGE.
  • Lyophilized from buffered solution; may contain trehalose for stability.
  • Store at -20°C; reconstituted stable at 4°C for 1 week or frozen for long-term storage.
  • Bioactivity ED50 ~1.0-2.4 ng/mL in cell proliferation assays.

Catalog No. 50-001-75958


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.