Learn More
Enzo Life Sciences M-CSF (mouse), (recombinant) (10µg)
SDP
Catalog No. 502013935
Encompass_Preferred
Alternative name: Macrophage colony stimulating factor, CSF-1. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized from 5mM sodium phosphate, pH 8.0, containing 100 mM NaCl. Source: Produced in E.coli. Long Term Storage: -20°C.
Catalog No. 50-201-3935 Supplier Enzo Life Sciences Supplier No. ENZPRT1440010
Alternative name: Macrophage colony stimulating factor, CSF-1. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized from 5mM sodium phosphate, pH 8.0, containing 100 mM NaCl. Source: Produced in E.coli. Long Term Storage: -20°C.