Learn More
Sino Biological Human M-CSF / CSF-1 Protein, Biotinylated 100ug
Supplier:  Sino Biological 11792HNAHB100
Encompass_Preferred
A DNA sequence encoding the human CSF1 (NP_757349.1) (Met1-Asn190) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6201