Learn More
Medchemexpress LLC M-CSF Rat 1mg
SDP
Catalog No. 5000179758
Encompass_Preferred
M-CSF Protein Rat is a pro-inflammatory cytokine binds to its receptor CSF1R and is involved in the development and proliferation of cells of the monocyte/macrophage lineage and participates in the induction of osteoclasts which are important in the destruction of bone and cartilage and in the periarticular osteoporotic changes
Catalog No. 50-001-79758 Supplier Medchemexpress LLC Supplier No. HYP73861MG
M-CSF Protein Rat is a pro-inflammatory cytokine binds to its receptor CSF1R and is involved in the development and proliferation of cells of the monocyte/macrophage lineage and participates in the induction of osteoclasts which are important in the destruction of bone and cartilage and in the periarticular osteoporotic changes