Learn More
Medchemexpress LLC M-CSF Rat HEK293 10ug | 10ug
SDP
Catalog No. 5000185006
Encompass_Preferred
The M-CSF protein regulates hematopoietic precursor cells promoting their survival proliferation and differentiation M-CSF Protein Rat (HEK293) is the recombinant rat-derived M-CSF protein expressed by HEK293 with tag free
Catalog No. 50-001-85006 Supplier Medchemexpress LLC Supplier No. HYP7049310UG
The M-CSF protein regulates hematopoietic precursor cells promoting their survival proliferation and differentiation M-CSF Protein Rat (HEK293) is the recombinant rat-derived M-CSF protein expressed by HEK293 with tag free