Learn More
ACROBiosystems ACROBiosystems Cynomolgus M-CSF R / CSF1R / CD115 Protein, Fc Tag
Supplier:  ACROBiosystems CSRC52521MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus M-CSF R, Fc Tag (CSR-C5252) is expressed from human 293 cells (HEK293). It contains AA Ile 99 - Pro 596 (Accession # G7P8P1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 81.9 kDa. As a result of glycosylation, the protein migrates as 110-125 kDa under reducing (R) condition, and 200- 240 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Biotinylated Human M-CSF, His,Avitag (Cat. No. MCF-H82E6) at 2 μg/mL (100 μL/well) on streptavidin precoated (0.2 μg/well) plate, can bind Cynomolgus M-CSF R, Fc Tag (Cat. No. CSR-C5252) with a linear range of 1-39 ng/mL (QC tested).;Immobilized Human M-CSF, His Tag (Cat. No. MCF-H5247) at 5 μg/mL (100 μL/well) can bind Cynomolgus M-CSF R, Fc Tag (Cat. No. CSR-C5252) with a linear range of 1-39 ng/mL (Routinely tested).
Catalog No. 50-195-7409