Learn More
ACROBiosystems ACROBiosystems Human M-CSF R / CSF1R / CD115 Protein, Fc Tag, low endotoxin
Supplier:  ACROBiosystems CSRH5258200UG
Encompass_Preferred
Recombinant Protein;200UG;Human M-CSF R, Fc Tag, low endotoxin (CSR-H5258) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Glu 512 (Accession # NP_005202.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 80.6 kDa. The protein migrates as 100-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human M-CSF, His Tag (Cat. No. MCF-H5247) at 5 μg/mL (100 μL/well) can bind Human M-CSF R, Fc Tag, low endotoxin (Cat. No. CSR-H5258) with a linear range of 0.1-4 ng/mL (QC tested).
Catalog No. 50-195-6750