Learn More
ACROBiosystems ACROBiosystems Rabbit M-CSF R / CSF1R / CD115 Protein, His Tag
Catalog No. 501956931
Encompass_Preferred
Recombinant Protein;1MG;Rabbit M-CSF R, His Tag (CSR-R82H5) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Ser 503 (Accession # G1TTE0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.2 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6931 Supplier ACROBiosystems Supplier No. CSRR82H51MG
Recombinant Protein;1MG;Rabbit M-CSF R, His Tag (CSR-R82H5) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Ser 503 (Accession # G1TTE0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.2 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.