Learn More
ACROBiosystems ACROBiosystems Canine M-CSF R / CSF1R / CD115 Protein, His Tag
Supplier:  ACROBiosystems CSRC82H31MG
Encompass_Preferred
Recombinant Protein;1MG;Canine M-CSF R, His Tag (CSR-C82H3) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 510 (Accession # E2RJV2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 56.1 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Canine M-CSF R, His Tag (Cat. No. CSR-C82H3) at 2 μg/mL (100 μL/well) can bind Biotinylated Human M-CSF, His Tag, Avi Tag (Cat. No. MCF-H82E6) with a linear range of 1-16 ng/mL (QC tested).
Catalog No. 50-195-6934