Learn More
ACROBiosystems ACROBiosystems Cynomolgus M-CSF R / CSF1R / CD115 Protein, His Tag
Catalog No. 501957404
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus M-CSF R, His Tag (CSR-C52E1) is expressed from human 293 cells (HEK293). It contains AA Ile 99 - Pro 596 (Accession # G7P8P1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.1 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus M-CSF R, His Tag (Cat. No. CSR-C52E1) at 5 μg/mL (100 μL/well) can bind Biotinylated Human M-CSF, His,Avitag (Cat. No. MCF-H82E6) with a linear range of 2-16 ng/mL (QC tested).
Catalog No. 50-195-7404 Supplier ACROBiosystems Supplier No. CSRC52E11MG
Recombinant Protein;1MG;Cynomolgus M-CSF R, His Tag (CSR-C52E1) is expressed from human 293 cells (HEK293). It contains AA Ile 99 - Pro 596 (Accession # G7P8P1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.1 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus M-CSF R, His Tag (Cat. No. CSR-C52E1) at 5 μg/mL (100 μL/well) can bind Biotinylated Human M-CSF, His,Avitag (Cat. No. MCF-H82E6) with a linear range of 2-16 ng/mL (QC tested).