Learn More
ACROBiosystems ACROBiosystems Mouse M-CSF R / CSF1R / CD115 Protein, His Tag
Supplier:  ACROBiosystems CSRM52E7100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse M-CSF R, His Tag (CSR-M52E7) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Ser 511 (Accession # NP_001032948).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.1 kDa. The protein migrates as 66-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse M-CSF Protein at 5 μg/mL (100 μL/well) can bind Mouse M-CSF R, His Tag (Cat. No. CSR-M52E7) with a linear range of 0.078-0.625 μg/mL (Routinely tested).
Catalog No. 50-195-7360