Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich m-Cymene

Catalog No. 255289500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-528-9500MG 500 mg
25-528-9100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-528-9500MG Supplier Sigma Aldrich Supplier No. 255289500MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

m-Cymene

Specifications

CAS 535-77-3
Melting Point -64°C (lit.)
Density 0.861 g/mL (at 25°C (literature))
Boiling Point 175°C (lit.)
Molecular Formula C10H14
Refractive Index n20/D 1.492 (literature)
Linear Formula CH3C6H4CH(CH3)2
MDL Number MFCD00008891
Quantity 500 mg
Synonym 1-Isopropyl-3-methylbenzene; 3-Isopropyltoluene
Molecular Weight (g/mol) 134.22
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.