Learn More
MSC SKILCRAFT Floor Pad, Nylon, Scrubbing, Green, 16" Diameter
Catalog No. 502839503
Encompass
These floor pads are designed for use on concentrated weight, rotary disk floor machines rated between 150-350 rpm. They are washable, reusable, and corrosion-resistant, making them suitable for various floor types including resilient tile, terrazzo, marble, and ceramic. Each pad has a 16 inch diameter, is 11/16 inch thick, and features a 3-3/8 inch center hole.
  • Penetrates deep into floor surfaces.
  • Resistant to wear.
  • Well-suited for a variety of tasks beyond standard scrubbing.
  • Effective with plain water.
  • Washable, reusable, and corrosion-resistant.
  • Suitable for resilient tile, terrazzo, marble, and ceramic floors.
Catalog No. 50-283-9503 Supplier MSC Supplier No. 89460471
These floor pads are designed for use on concentrated weight, rotary disk floor machines rated between 150-350 rpm. They are washable, reusable, and corrosion-resistant, making them suitable for various floor types including resilient tile, terrazzo, marble, and ceramic. Each pad has a 16 inch diameter, is 11/16 inch thick, and features a 3-3/8 inch center hole.
  • Penetrates deep into floor surfaces.
  • Resistant to wear.
  • Well-suited for a variety of tasks beyond standard scrubbing.
  • Effective with plain water.
  • Washable, reusable, and corrosion-resistant.
  • Suitable for resilient tile, terrazzo, marble, and ceramic floors.